Glutaconaldehydedianil hydrochloride mixture of isomers 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-(5-Anilino-2,4-pentadienylidene)aniline hydrochloride, N-[5-(Phenylamino)-2,4-pentadienylidene]aniline monohydrochloride, N-((2E,4E)-5-(Phenylamino)penta-2,4-dien-1-ylidene)aniline hydrochloride
A18390-22
A18390-06
A18390-14
CAAAAA18390-22EA
1540.5
CAD
CAAAAA18390-22
CAAAA18390-06
CAAAA18390-14
Glutaconaldehydedianil hydrochloride mixture of isomers 98%
Glutaconaldehydedianil hydrochloride
Purity: 98%
CAS No.: 1497-49-0
Molecular Formula: C6H5NHCH=CHCH=CHCH=NC6H5.HCl
Formula Weight: 284.79
Melting Point: ca 124-132°
Beilstein Reference: 3916924
MDL No.: MFCD00012630
Formula:
C₁₇H₁₆N₂ MW: 284.79 g/mol Melting Pt: ∼ 124…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012630 CAS Number: 1497-49-0 EINECS: 216-094-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...