4-Acetyldiphenyl sulfone 98%

Supplier: Thermo Scientific Chemicals

A18376-03 A18376-06 A18376-14
CAAAA18376-03EA 150.1 CAD
CAAAA18376-03 CAAAA18376-06 CAAAA18376-14
4-Acetyldiphenyl sulfone 98%
4-Acetyldiphenyl sulfone

Purity: 98% CAS No.: 65085-83-8 Molecular Formula: C14H12O3S Formula Weight: 260.31 Melting Point: 138-140° MDL No.: MFCD00209628

Formula: C₁₄H₁₂O₃S
Melting Pt: 138…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00209628
CAS Number: 65085-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR