1-(2-Aminoethyl)piperidine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-(2-Aminoethyl)piperidine

A18373-06 A18373-14
CAAAA18373-06EA 76.95 CAD
CAAAA18373-06 CAAAA18373-14
1-(2-Aminoethyl)piperidine 98%
1-(2-Aminoethyl)piperidine

Purity: 98% CAS No.: 27578-60-5 Molecular Formula: C7H16N2 Formula Weight: 128.22 Flash Point: 57°(134°F) Refractive Index: 1.4740 Boiling Point: 185-186° Beilstein Reference: 103469 UN No.: 2733 MDL No.: MFCD00006516

Formula: C₇H₁₆N₂
MW: 128.22 g/mol
Boiling Pt: 185…186 °C
Density: 0.903
Flash Pt: 57 °C(134 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006516
CAS Number: 27578-60-5
EINECS: 248-540-8
UN: 2924
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR