O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride ≥99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18368-06
A18368-01
A18368-03
CAAAAA18368-06EA
1248.2
CAD
CAAAAA18368-06
CAAAA18368-01
CAAAA18368-03
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride ≥99%
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride
Purity: 99+%
CAS No.: 57981-02-9
Molecular Formula: C7H4F5NO.HCl
Formula Weight: 249.57
Melting Point: ca 215° subl.
Beilstein Reference: 4031190
MDL No.: MFCD00012953
Formula:
C₇H₄F₅NO Melting Pt: ca 215 °C (subl.) Storage Temperature: Ambient |
MDL Number:
MFCD00012953 CAS Number: 57981-02-9 EINECS: 261-057-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...