L-Tyrosine ethyl ester hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: H-Tyr-OEt·HCl, H-Tyr-OEt.HCl

A18358-14 A18358-22
CAAAAA18358-14EA 115.81 CAD
CAAAAA18358-14 CAAAAA18358-22
L-Tyrosine ethyl ester hydrochloride 99%
L-Tyrosine ethyl ester hydrochloride

Purity: 99% CAS No.: 4089-07-0 Molecular Formula: C11H15NO3.HCl Formula Weight: 245.71 Melting Point: 166-168° Beilstein Reference: 4725904 Optical Rotation: -6.8° (c=2 in water) MDL No.: MFCD00063047

Formula: C₁₁H₁₅NO₃
MW: 245.70 g/mol
Melting Pt: 166…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00063047
CAS Number: 4089-07-0
EINECS: 223-820-2

Order Now

Specification Test Results

Optical rotation -6.8° (c=2 in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR