trans,cis-2,6-Nonadienal ≥90% remainder mainly trans,trans-isomer

Supplier: Thermo Scientific Chemicals
Warning

A18356-03 A18356-06 A18356-14
CAAAA18356-03EA 54.67 CAD
CAAAA18356-03 CAAAA18356-06 CAAAA18356-14
trans,cis-2,6-Nonadienal ≥90% remainder mainly trans,trans-isomer
trans,cis-2,6-Nonadienal

Purity: 90+% CAS No.: 557-48-2 Molecular Formula: C9H14O Formula Weight: 138.20 Flash Point: 76°(169°F) Refractive Index: 1.4740 Boiling Point: 90°/4mm MDL No.: MFCD00007009

Formula: C₉H₁₄O
MW: 138.21 g/mol
Boiling Pt: 90 °C (4 mmHg)
Density: 0.866
Flash Pt: 76 °C (169 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007009
CAS Number: 557-48-2
EINECS: 209-178-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR