(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18305-06
A18305-14
CAAAA18305-03EA
112.4
CAD
CAAAA18305-03
CAAAA18305-06
CAAAA18305-14
(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate 98%
(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate
Fieser: 5,90 6,93 8,76 10,78
Purity: 98%
CAS No.: 52186-89-7
Molecular Formula: C24H24BF4O2P
Formula Weight: 462.23
Melting Point: 173-179°
Beilstein Reference: 4650673
Formula:
C₂₄H₂₄BF₄O₂P Melting Pt: 173…179 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051879 CAS Number: 52186-89-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...