Indium(III) phosphate 98%

Supplier: Thermo Scientific Chemicals
Warning

A18281-06 A18281-14
CAAAAA18281-06EA 74.89 CAD
CAAAAA18281-06 CAAAAA18281-14
Indium(III) phosphate 98%

Purity: 98% CAS No.: 14693-82-4 Molecular Formula: InPO4 Formula Weight: 209.79 MDL No.: MFCD00016154

Formula: InPO₄
Density: 4.9
Storage Temperature: Ambient
MDL Number: MFCD00016154
CAS Number: 14693-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR