Acid green 1

Supplier: Thermo Scientific Chemicals

A18268-14 A18268-22
CAAAA18268-14EA 47.4 CAD
CAAAA18268-14 CAAAA18268-22
Acid green 1
Acid green 1

CAS No.: 19381-50-1 Molecular Formula: C30H15FeN3Na3O15S3 Formula Weight: 878.47 MDL No.: MFCD00003886

Formula: C₃₀H₁₈FeN₃O₁₅S₃
Storage Temperature: Ambient
MDL Number: MFCD00003886
CAS Number: 19381-50-1
EINECS: 243-010-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR