N-Acetyl-ε-caprolactam 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-Acetyl-2-oxohexamethyleneimine, N-Acetylcaprolactam
A18257-14
A18257-22
CAAAA18257-14EA
53.4
CAD
CAAAA18257-14
CAAAA18257-22
N-Acetyl-ε-caprolactam 99%
N-Acetyl-epsilon-caprolactam
Purity: 99%
CAS No.: 1888-91-1
Molecular Formula: C8H13NO2
Formula Weight: 155.20
Flash Point: 122°(252°F)
Refractive Index: 1.4890
Boiling Point: 124°/12mm
Beilstein Reference: 119598
MDL No.: MFCD00003262
Formula:
C₈H₁₃NO₂ MW: 155.20 g/mol Boiling Pt: 124 °C (12 mmHg) Density: 1.094 Flash Pt: 122 °C (252 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003262 CAS Number: 1888-91-1 EINECS: 217-565-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...