N-Carbobenzoxy-L-glutamine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Cbz-L-glutamine, Z-Gln-OH

A18240-06 A18240-14
CAAAA18240-06EA 37.29 CAD
CAAAA18240-06 CAAAA18240-14
N-Carbobenzoxy-L-glutamine 99%
N-Carbobenzoxy-L-glutamine

Purity: 99% CAS No.: 2650-64-8 Molecular Formula: C13H16N2O5 Formula Weight: 280.28 Melting Point: 134-137° Beilstein Reference: 2061271 Optical Rotation: -7° (c=2 in ethanol) MDL No.: MFCD00008043

Formula: C₁₃H₁₆N₂O₅
MW: 280.28 g/mol
Melting Pt: 134…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00008043
CAS Number: 2650-64-8
EINECS: 220-173-8

Order Now

Specification Test Results

Optical rotation -7° (c=2 in ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR