3-(4-Methoxybenzoyl)propionic acid 98+%
Supplier: Thermo Scientific Chemicals
A18210-06
A18210-14
CAAAAA18210-22EA
717.46
CAD
CAAAAA18210-22
CAAAA18210-06
CAAAA18210-14
3-(4-Methoxybenzoyl)propionic acid 98+%
3-(4-Methoxybenzoyl)propionic acid
Purity: 98+%
CAS No.: 3153-44-4
Molecular Formula: C11H12O4
Formula Weight: 208.21
Melting Point: 147-151°
Beilstein Reference: 785074
Formula:
C₁₁H₁₂O₄ Melting Pt: 147…151 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002795 CAS Number: 3153-44-4 EINECS: 221-593-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...