(S)-2-Hydroxy-3-phenylpropanoic acid 98%
Supplier: Thermo Scientific Chemicals
A18203-03
A18203-06
A18203-14
CAAAA18203-03EA
46.45
CAD
CAAAA18203-03
CAAAA18203-06
CAAAA18203-14
(S)-2-Hydroxy-3-phenylpropanoic acid 98%
(S)-2-Hydroxy-3-phenylpropanoic acid
Purity: 98%
CAS No.: 20312-36-1
Molecular Formula: C9H10O3
Formula Weight: 166.18
Storage Temperature: 2°C to 8°C
Melting Point: 122-126°
Beilstein Reference: 2209792
Optical Rotation: -24° (c=1 in water)
MDL No.: MFCD00004244
Formula:
C₉H₁₀O₃ Melting Pt: 122…126 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004244 CAS Number: 20312-36-1 EINECS: 243-726-5 |
Specification Test Results
Optical rotation | -22° (c=1 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...