Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: Bz-Arg-OEt.HCl, Nα-Benzoyl-L-arginine Ethyl Ester HCl

A18181-03 A18181-06
CAAAA18181-03EA 40.92 CAD
CAAAA18181-03 CAAAA18181-06
Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride ≥98%
Nalpha-Benzoyl-L-arginine ethyl ester hydrochloride

Purity: 98+% CAS No.: 2645-08-1 Molecular Formula: C15H22N4O3.HCl Formula Weight: 342.84 Storage Temperature: -30°C to -10°C Melting Point: 127-131° Beilstein Reference: 3781694 Optical Rotation: -17° (c=2, water) MDL No.: MFCD00012579

Formula: C₁₅H₂₂N₄O₃
Melting Pt: 127…131 °C
Storage Temperature: Freezer
MDL Number: MFCD00012579
CAS Number: 2645-08-1
EINECS: 220-157-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR