5-Nitro-2-furaldehyde diacetate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Nitrofurfural Diacetyl Acetal, Nitrofurfural diacetate, 2-(Diacetoxymethyl)-5-nitrofuran, (5-Nitrofuran-2-yl)methylene diacetate, 5-Nitrofurfurylidene Diacetate

A18169-22 A18169-36
CAAAAA18169-0EEA 883.73 CAD
CAAAAA18169-0E CAAAA18169-22 CAAAA18169-36
5-Nitro-2-furaldehyde diacetate 97%
5-Nitro-2-furaldehyde diacetate

Purity: 98+% CAS No.: 92-55-7 Molecular Formula: C9H9NO7 Formula Weight: 243.17 Melting Point: 87-92° Beilstein Reference: 255089 MDL No.: MFCD00003244

Formula: C₉H₉NO₇
MW: 243.17 g/mol
Melting Pt: 87…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00003244
CAS Number: 92-55-7
EINECS: 202-166-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR