1-Allyl-3-(2-hydroxyethyl)-2-thiourea 97%

Supplier: Thermo Scientific Chemicals
Warning

A18161-18 A18161-30
CAAAA18161-18EA 69.05 CAD
CAAAA18161-18 CAAAA18161-30
1-Allyl-3-(2-hydroxyethyl)-2-thiourea 97%
1-Allyl-3-(2-hydroxyethyl)-2-thiourea

Purity: 97% CAS No.: 105-81-7 Molecular Formula: C6H12N2OS Formula Weight: 160.24 Melting Point: 77-79° MDL No.: MFCD00002838

Formula: C₆H₁₂N₂OS
Melting Pt: 77…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00002838
CAS Number: 105-81-7
EINECS: 203-334-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR