L-Lysine methyl ester dihydrochloride 99%
Supplier: Thermo Scientific Chemicals
A18157-14
A18157-22
A18157-36
CAAAA18157-14EA
96.7
CAD
CAAAA18157-14
CAAAA18157-22
CAAAA18157-36
L-Lysine methyl ester dihydrochloride 99%
L-Lysine methyl ester dihydrochloride
Purity: 99%
CAS No.: 26348-70-9
Molecular Formula: C7H16N2O2.2HCl
Formula Weight: 233.14
Storage Temperature: 2°C to 8°C
Melting Point: ca 200° dec.
Beilstein Reference: 5302183
MDL No.: MFCD00039067
Formula:
C₇H₁₆N₂O₂ Melting Pt: ca 200 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00039067 CAS Number: 26348-70-9 EINECS: 247-625-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...