2-Bromobenzylamine hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

A18151-06 A18151-14
CAAAA18151-06EA 120.71 CAD
CAAAA18151-06 CAAAA18151-14 CAAAA18151-22
2-Bromobenzylamine hydrochloride 97%
2-Bromobenzylamine hydrochloride

Purity: 97% CAS No.: 5465-63-4 Molecular Formula: C7H8BrN.HCl Formula Weight: 222.52 Melting Point: 227-230° Beilstein Reference: 3913456

Formula: C₇H₉BrClN
Melting Pt: 227…230 °C
Storage Temperature: Ambient
MDL Number: MFCD00012853
CAS Number: 5465-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR