Nigrosine, water soluble

Supplier: Thermo Scientific Chemicals

A18147-14 A18147-22
CAAAA18147-14EA 44.56 CAD
CAAAA18147-14 CAAAA18147-22
Nigrosine, water soluble
Nigrosine

Fairly common biological stain for tissues and cells


CAS No.: 8005-03-6 Physical Form: Powder MDL No.: MFCD00044681


Applications: Fairly common biological stain for tissues and cells

Storage Temperature: Ambient
MDL Number: MFCD00044681
CAS Number: 8005-03-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR