6-Nitropiperonyl alcohol ≥98%
Supplier: Thermo Scientific Chemicals
A18140-06
A18140-14
CAAAA18140-06EA
192.76
CAD
CAAAA18140-06
CAAAA18140-14
6-Nitropiperonyl alcohol ≥98%
6-Nitropiperonyl alcohol
Purity: 98+%
CAS No.: 15341-08-9
Molecular Formula: C8H7NO5
Formula Weight: 197.15
Melting Point: 121-125°
Beilstein Reference: 13749
MDL No.: MFCD00005825
Formula:
C₈H₇NO₅ Melting Pt: 121…125 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005825 CAS Number: 15341-08-9 EINECS: 239-376-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...