2-Naphthyl phenyl ketone 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
2-Benzoylnaphthalene
A18121-03
A18121-06
CAAAA18121-03EA
76.95
CAD
CAAAA18121-03
CAAAA18121-06
2-Naphthyl phenyl ketone 98%
2-Naphthyl phenyl ketone
Purity: 98%
CAS No.: 644-13-3
Molecular Formula: C17H12O
Formula Weight: 232.28
Melting Point: 80-82°
Boiling Point: 216-216°/7mm
MDL No.: MFCD00004106
Formula:
C₁₇H₁₂O MW: 232.28 g/mol Boiling Pt: 216…216 °C/7 mm Melting Pt: 80…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004106 CAS Number: 644-13-3 EINECS: 211-410-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...