4-(2-Pyridylazo)resorcinol monosodim salt monohydrate

Supplier: Thermo Scientific Chemicals
Warning

A18103-03 A18103-06 A18103-14
CAAAAA18103-03EA 48.19 CAD
CAAAAA18103-03 CAAAAA18103-06 CAAAAA18103-14
4-(2-Pyridylazo)resorcinol monosodim salt monohydrate
4-(2-Pyridylazo)resorcinol monosodim salt monohydrate

CAS No.: 16593-81-0 Molecular Formula: C11H8N3NaO2.H2O Formula Weight: 255.21 (237.19anhy) Melting Point: 300° Beilstein Reference: 3825637 MDL No.: MFCD00150712

Formula: C₁₁H₈N₃NaO₂·1H₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150712
CAS Number: 16593-81-0
EINECS: 236-339-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR