Hydrocortisone-21-acetate 97+%

Supplier: Thermo Scientific Chemicals
Danger

A18089-03 A18089-06
CAAAA18089-03EA 69.52 CAD
CAAAA18089-03 CAAAA18089-06
Hydrocortisone-21-acetate 97+%
Hydrocortisone 21-acetate

Purity: 97+% CAS No.: 50-03-3 Molecular Formula: C23H32O6 Formula Weight: 404.51 Melting Point: ca 220° dec. Beilstein Reference: 2066841 Merck Reference: 14,4787 MDL No.: MFCD00037714

Formula: C₂₃H₃₂O₆
Melting Pt: ca 220 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00037714
CAS Number: 50-03-3
EINECS: 200-004-4
Merck Index: 14,4787

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR