DL-Methionine sulfoxide 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Amino-4-(methylsulfinyl)butanoic acid, 2-Amino-4-(methylsulphinyl)butanoic acid

A18081-06 A18081-14
CAAAA18081-06EA 105.7 CAD
CAAAA18081-06 CAAAA18081-14
DL-Methionine sulfoxide 98+%
DL-Methionine sulphoxide

Purity: 98+% CAS No.: 62697-73-8 Molecular Formula: C5H11NO3S Formula Weight: 165.21 Melting Point: ca 230° dec. MDL No.: MFCD00002620

Formula: C₅H₁₁NO₃S
MW: 165.21 g/mol
Melting Pt: ca 230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002620
CAS Number: 62697-73-8
EINECS: 263-700-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR