4-Hydroxy-3-methoxybenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

A18056-14 A18056-22
CAAAA18056-06EA 76.69 CAD
CAAAA18056-06 CAAAA18056-14 CAAAA18056-22
4-Hydroxy-3-methoxybenzonitrile 98%
4-Hydroxy-3-methoxybenzonitrile

Purity: 98% CAS No.: 4421-08-3 Molecular Formula: C8H7NO2 Formula Weight: 149.15 Melting Point: 85-89° UN No.: 3439 MDL No.: MFCD00001820

Formula: C₈H₇NO₂
MW: 149.15 g/mol
Melting Pt: 85…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00001820
CAS Number: 4421-08-3
EINECS: 224-589-0
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR