Methyl-4-hydroxy-3,5-dimethoxybenzoate ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18054-06
A18054-14
CAAAAA18054-06EA
85.64
CAD
CAAAAA18054-06
CAAAA18054-14
CAAAA18054-22
Methyl-4-hydroxy-3,5-dimethoxybenzoate ≥98%
Methyl-4-hydroxy-3,5-dimethoxybenzoate
Purity: 98+%
CAS No.: 884-35-5
Molecular Formula: C10H12O5
Formula Weight: 212.20
Melting Point: 102-108°
Beilstein Reference: 2217524
MDL No.: MFCD00017199
Formula:
C₁₀H₁₂O₅ Melting Pt: 102…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017199 CAS Number: 884-35-5 EINECS: 429-050-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...