2-Acetyl-6-methoxynaphthalene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6'-Methoxy-2'-acetonaphthone

A18031-14 A18031-22
CAAAA18031-14EA 230.68 CAD
CAAAA18031-14 CAAAA18031-22 CAAAA18031-36
2-Acetyl-6-methoxynaphthalene 98%
2-Acetyl-6-methoxynaphthalene

Purity: 98% CAS No.: 3900-45-6 Molecular Formula: C13H12O2 Formula Weight: 200.24 Melting Point: 107-109° Boiling Point: 155-160°/0.4mm Beilstein Reference: 1871098 MDL No.: MFCD00021643

Formula: C₁₃H₁₂O₂
MW: 200.24 g/mol
Boiling Pt: 155…160 °C (0.4 mmHg)
Melting Pt: 107…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00021643
CAS Number: 3900-45-6
EINECS: 223-453-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR