Monosodium 5-sulfoisophthalate 95%

Supplier: Thermo Scientific Chemicals
Warning

A18028-22 A18028-36 A18028-30
CAAAAA18028-22EA 58.93 CAD
CAAAAA18028-22 CAAAAA18028-36 CAAAA18028-30
Monosodium 5-sulfoisophthalate 95%
Monosodium 5-sulfoisophthalate

Purity: 95% CAS No.: 6362-79-4 Molecular Formula: C8H5NaO7S Formula Weight: 268.18 Melting Point: 300° Beilstein Reference: 4170770 MDL No.: MFCD00007495

Formula: C₈H₆O₇S
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007495
CAS Number: 6362-79-4
EINECS: 228-845-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR