Amberlyst® A-21, Amberlyst™ ion exchange resin

Supplier: Thermo Scientific Chemicals

A17956-0B A17956-30
CAAAA17956-0BEA 420.28 CAD
CAAAA17956-0B CAAAA17956-30
Amberlyst® A-21, Amberlyst™ ion exchange resin
Amberlyst® A-21

Fieser: 18,194 21,237


CAS No.: 9049-93-8 Odor: Odorless MDL No.: MFCD00145842

Storage Temperature: Ambient
MDL Number: MFCD00145842
CAS Number: 9049-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR