Methyl (4-methoxyphenyl)acetate 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17940-14
A17940-22
A17940-36
CAAAA17940-14EA
71.89
CAD
CAAAA17940-14
CAAAA17940-22
CAAAA17940-36
Methyl (4-methoxyphenyl)acetate 97+%
Methyl (4-methoxyphenyl)acetate
Purity: 97+%
CAS No.: 23786-14-3
Molecular Formula: C10H12O3
Formula Weight: 180.23
Flash Point:
Formula:
C₁₀H₁₂O₃ Boiling Pt: 157…159 °C/19 mm Density: 1.135 Flash Pt: >100 °C(212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008454 CAS Number: 23786-14-3 EINECS: 245-886-1 UN: 3272 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...