5-Bromo-1,3-benzodioxole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromo-1,2-(methylenedioxy)benzene, 5-Bromobenzo[d][1,3]dioxole

A17906-06 A17906-36 A17906-14 A17906-22
CAAAAA17906-06EA 54.04 CAD
CAAAAA17906-06 CAAAAA17906-36 CAAAA17906-14 CAAAA17906-22
5-Bromo-1,3-benzodioxole 98%
5-Bromo-1,3-benzodioxole

Purity: 98% CAS No.: 2635-13-4 Molecular Formula: C7H5BrO2 Formula Weight: 201.02 Flash Point: 110°(230°F) Refractive Index: 1.5840 Boiling Point: 228° Beilstein Reference: 127407 MDL No.: MFCD00005821

Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Boiling Pt: 228 °C
Density: 1.669
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005821
CAS Number: 2635-13-4
EINECS: 220-123-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR