Methyl glycolate 98%
Supplier: Thermo Scientific Chemicals
Warning
|
A17870-22
A17870-06
A17870-14
CAAAAA17870-22EA
518.24
CAD
CAAAAA17870-22
CAAAA17870-06
CAAAA17870-14
Methyl glycolate 98%
Methyl glycolate
Purity: 98%
CAS No.: 96-35-5
Molecular Formula: C3H6O3
Formula Weight: 90.08
Flash Point: 67°(152°F)
Refractive Index: 1.4170
Boiling Point: 150-151°
Beilstein Reference: 1699571
MDL No.: MFCD00004667
Formula:
C₃H₆O₃ Boiling Pt: 150…151 °C Density: 1.184 Flash Pt: 67 °C(152 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004667 CAS Number: 96-35-5 EINECS: 202-502-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...