Manganese(III) phosphate hydrate 99%
Supplier: Thermo Scientific Chemicals
A17868-09
A17868-18
CAAAAA17868-09EA
78.53
CAD
CAAAAA17868-09
CAAAAA17868-18
Manganese(III) phosphate hydrate 99%
Manganese(III) phosphate hydrate
Purity: 99%
CAS No.: 104663-56-1
Molecular Formula: MnPO4.xH2O
Formula Weight: 149.91(anhy)
MDL No.: MFCD00209658
Formula:
MnPO₄·xH₂O MW: 149.91 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00209658 CAS Number: 104663-56-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...