L-Isoleucine methyl ester hydrochloride ≥98%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-Ile-OMe·HCl, H-Ile-OMe.HCl
A17847-03
A17847-06
A17847-14
CAAAA17847-03EA
63.04
CAD
CAAAA17847-03
CAAAA17847-06
CAAAA17847-14
L-Isoleucine methyl ester hydrochloride ≥98%
L-Isoleucine methyl ester hydrochloride
Purity: 98+%
CAS No.: 18598-74-8
Molecular Formula: C7H15NO2.HCl
Formula Weight: 181.66
Melting Point: 98-100°
Beilstein Reference: 3595132
MDL No.: MFCD00038911
Formula:
C₇H₁₅NO₂ MW: 181.66 g/mol Melting Pt: 98…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038911 CAS Number: 18598-74-8 EINECS: 242-437-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...