Benzo[h]quinoline 98%

Supplier: Thermo Scientific Chemicals
Warning

A17794-03 A17794-06
CAAAA17794-03EA 50.09 CAD
CAAAA17794-03 CAAAA17794-06
Benzo[h]quinoline 98%
Benzo[h]quinoline

Purity: 98% CAS No.: 230-27-3 Molecular Formula: C13H9N Formula Weight: 179.22 Flash Point: 110°(230°F) Melting Point: 48-52° Boiling Point: 233°/47mm Beilstein Reference: 120249 MDL No.: MFCD00004984

Formula: C₁₃H₉N
Boiling Pt: 233 °C/47 mm
Melting Pt: 48…52 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004984
CAS Number: 230-27-3
EINECS: 205-937-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR