4-Bromo-2,2-diphenylbutanenitrile 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Bromo-2,2-diphenylbutyronitrile
A17786-22
A17786-36
CAAAA17786-14EA
127.81
CAD
CAAAA17786-14
CAAAA17786-22
CAAAA17786-36
4-Bromo-2,2-diphenylbutanenitrile 95%
4-Bromo-2,2-diphenylbutanenitrile
Purity: 95%
CAS No.: 39186-58-8
Molecular Formula: C16H14BrN
Formula Weight: 300.20
Melting Point: 67-69°
Beilstein Reference: 1684952
UN No.: 3439
MDL No.: MFCD00001845
Formula:
C₁₆H₁₄BrN MW: 300.20 g/mol Melting Pt: 67…69 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001845 CAS Number: 39186-58-8 EINECS: 254-337-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...