1-Pyrenebutyric acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17760-03
A17760-06
CAAAA17760-03EA
83.42
CAD
CAAAA17760-03
CAAAA17760-06
1-Pyrenebutyric acid 97%
1-Pyrenebutyric acid
Purity: 97%
CAS No.: 3443-45-6
Molecular Formula: C20H16O2
Formula Weight: 288.35
Melting Point: 184-186°
Beilstein Reference: 2140554
Formula:
C₂₀H₁₆O₂ Melting Pt: 184…186 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004141 CAS Number: 3443-45-6 EINECS: 222-354-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...