2,2':6',2''-Terpyridine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17750-01
A17750-03
CAAAA17750-01EA
97.17
CAD
CAAAA17750-01
CAAAA17750-03
2,2':6',2''-Terpyridine 97%
2,2':6',2''-Terpyridine
Purity: 97%
CAS No.: 1148-79-4
Molecular Formula: C15H11N3
Formula Weight: 233.28
Physical Form: Powder
Odor: Odorless
UN No.: 2811
MDL No.: MFCD00006213
Formula:
C₁₅H₁₁N₃ Melting Pt: 89…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006213 CAS Number: 1148-79-4 EINECS: 214-559-5 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...