Amberlite® IRA-410 (Cl), Amberlite™ ion exchange resin

Supplier: Thermo Scientific Chemicals

Synonyms: Amberlite® IRA-410 Cl type

A17734-0E A17734-22 A17734-36
CAAAA17734-0EEA 448.72 CAD
CAAAA17734-0E CAAAA17734-22 CAAAA17734-36
Amberlite® IRA-410 (Cl), Amberlite™ ion exchange resin
Amberlite® IRA-410 (Cl)

Styrene-DVB; Strongly basic anion exchange resin.


CAS No.: 9002-26-0 Odor: Amine-like MDL No.: MFCD00132710

Storage Temperature: Ambient
MDL Number: MFCD00132710
CAS Number: 9002-26-0
Merck Index: 14,382

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR