DL-Glutamic acid monohydrate 99%

Supplier: Thermo Scientific Chemicals

A17719-14 A17719-22 A17719-36
CAAAAA17719-14EA 48.35 CAD
CAAAAA17719-14 CAAAAA17719-22 CAAAAA17719-36
DL-Glutamic acid monohydrate 99%
DL-Glutamic acid monohydrate

Purity: 99% CAS No.: 19285-83-7 Molecular Formula: C5H9NO4.H2O Formula Weight: 165.15 (147.13anhy) Melting Point: ca 200° dec. Beilstein Reference: 4155097 Merck Reference: 14,4469 MDL No.: MFCD00150703

Formula: C₅H₉NO₄·1H₂O
Melting Pt: ca 200 °C (dec.)
Density: 1.460
Storage Temperature: Ambient
MDL Number: MFCD00150703
CAS Number: 19285-83-7
EINECS: 210-522-2
Merck Index: 14,4469

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR