(S)-2-(((Benzyloxy)carbonyl)amino)pentanedioic acid 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Cbz-Glu-OH, Z-Glu-OH, N-Cbz-L-glutamic Acid

A17686-09 A17686-18
CAAAA17686-09EA 63.83 CAD
CAAAA17686-09 CAAAA17686-18
(S)-2-(((Benzyloxy)carbonyl)amino)pentanedioic acid 99%
(S)-2-(((Benzyloxy)carbonyl)amino)pentanedioic acid

Purity: 99% CAS No.: 1155-62-0 Molecular Formula: C13H15NO6 Formula Weight: 281.27 Melting Point: 108-116° Beilstein Reference: 2061272 Optical Rotation: -7.5° (c=4 in acetic acid) MDL No.: MFCD00002801

Formula: C13H15NO6
Melting Pt: 108…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00002801
CAS Number: 1155-62-0
EINECS: 214-584-1

Order Now

Specification Test Results

Optical rotation -7.5° (c=4 in acetic acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR