4-(4-Fluorobenzyloxy)benzaldehyde 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-((4-Fluorobenzyl)oxy)benzaldehyde

A17657-14 A17657-06
CAAAAA17657-14EA 219.62 CAD
CAAAAA17657-14 CAAAA17657-06
4-(4-Fluorobenzyloxy)benzaldehyde 97%
4-(4-Fluorobenzyloxy)benzaldehyde

Purity: 97% CAS No.: 56442-17-2 Molecular Formula: C14H11FO2 Formula Weight: 230.24 MDL No.: MFCD00173650

Formula: C₁₄H₁₁FO₂
MW: 230.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173650
CAS Number: 56442-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR