4-Hydroxybenzylideneacetone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxybenzilideneacetone, 4-(4-Hydroxyphenyl)-3-buten-2-one, 4-(4-Hydroxyphenyl)but-3-en-2-one, 4-Hydroxybenzalacetone

A17605-14 A17605-22
CAAAA17605-14EA 125.45 CAD
CAAAA17605-14 CAAAA17605-22 CAAAA17605-36
4-Hydroxybenzylideneacetone 97%
4-Hydroxybenzylideneacetone

Purity: 97% CAS No.: 3160-35-8 Molecular Formula: C10H10O2 Formula Weight: 162.19 Melting Point: 106-111° Beilstein Reference: 2412544

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Melting Pt: 106…111 °C
Storage Temperature: Ambient
MDL Number: MFCD00016490
CAS Number: 3160-35-8
EINECS: 221-607-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR