Ethyl (4-nitrobenzoyl)acetate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-(4-Nitrophenyl)-3-oxopropionic Acid Ethyl Ester, (4-Nitrobenzoyl)acetic Acid Ethyl Ester, Ethyl 3-(4-Nitrophenyl)-3-oxopropionate

A17603-09 A17603-18 A17603-30
CAAAA17603-09EA 102.7 CAD
CAAAA17603-09 CAAAA17603-18 CAAAA17603-30
Ethyl (4-nitrobenzoyl)acetate 97%
Ethyl (4-nitrobenzoyl)acetate

Purity: 97% CAS No.: 838-57-3 Molecular Formula: C11H11NO5 Formula Weight: 237.21 Melting Point: 68-73° Beilstein Reference: 1124763 MDL No.: MFCD00007357

Formula: C₁₁H₁₁NO₅
MW: 237.21 g/mol
Melting Pt: 68…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00007357
CAS Number: 838-57-3
EINECS: 212-656-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR