N-Carbobenzoxy-L-threonine 99%
Supplier: Thermo Scientific Chemicals
A17565-06
A17565-14
CAAAA17565-06EA
56.09
CAD
CAAAA17565-06
CAAAA17565-14
N-Carbobenzoxy-L-threonine 99%
N-Carbobenzoxy-L-threonine
Purity: 99%
CAS No.: 19728-63-3
Molecular Formula: C12H15NO5
Formula Weight: 253.26
Melting Point: 103-105°
Beilstein Reference: 2335409
Optical Rotation: -4.5° (c=4 in acetic acid)
MDL No.: MFCD00065948
Formula:
C₁₂H₁₅NO₅ Melting Pt: 103…105 °C Storage Temperature: Ambient |
MDL Number:
MFCD00065948 CAS Number: 19728-63-3 EINECS: 243-258-1 |
Specification Test Results
Optical rotation | -4.5° (c=4 in acetic acid) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...