Isopropyl-4-aminobenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Isopropyl 4-aminobenzoate

A17551-14 A17551-22
CAAAA17551-14EA 79 CAD
CAAAA17551-14 CAAAA17551-22
Isopropyl-4-aminobenzoate 98%
Isopropyl 4-aminobenzoate

Purity: 98% CAS No.: 18144-43-9 Molecular Formula: C10H13NO2 Formula Weight: 179.21 MDL No.: MFCD00017108

Formula: C₁₀H₁₃NO₂
MW: 179.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017108
CAS Number: 18144-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR