Isosorbide dimethyl ether 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(3R,3aR,6S,6aR)-3,6-Dimethoxyhexahydrofuro[3,2-b]furan
A17516-22
A17516-14
CAAAAA17516-22EA
279.66
CAD
CAAAAA17516-22
CAAAA17516-14
Isosorbide dimethyl ether 98%
Isosorbide dimethyl ether
Purity: 98%
CAS No.: 5306-85-4
Molecular Formula: C8H14O4
Formula Weight: 174.20
Flash Point: 108°(226°F)
Refractive Index: 1.4620
Melting Point: -70°
Boiling Point: 93-95°/0.1mm
Beilstein Reference: 80854
MDL No.: MFCD00064828
Formula:
C₈H₁₄O₄ MW: 174.20 g/mol Boiling Pt: 93…95 °C/0.1 mm Melting Pt: –70 °C Density: 1.165 Flash Pt: 108 °C(226 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064828 CAS Number: 5306-85-4 EINECS: 226-159-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...