N,O-Dimethylhydroxylamine hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17469-06
A17469-14
A17469-22
CAAAA17469-06EA
76.95
CAD
CAAAA17469-06
CAAAA17469-14
CAAAA17469-22
N,O-Dimethylhydroxylamine hydrochloride 98%
N,O-Dimethylhydroxylamine hydrochloride
Fieser: 11,201 12,205 15,144
Purity: 98%
CAS No.: 6638-79-5
Molecular Formula: CH3ONHCH3.HCl
Formula Weight: 97.55
Melting Point: 112-115°
Beilstein Reference: 3650353
Formula:
C₂H₈ClNO Melting Pt: 112…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012485 CAS Number: 6638-79-5 EINECS: 229-642-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...