N-tert-Butyl-ɑ-phenylnitrone 98%
Supplier: Thermo Scientific Chemicals
A17442-03
A17442-06
CAAAA17442-03EA
120.4
CAD
CAAAA17442-03
CAAAA17442-06
N-tert-Butyl-ɑ-phenylnitrone 98%
N-tert-Butyl-alpha-phenylnitrone
Purity: 98%
CAS No.: 3376-24-7
Molecular Formula: C11H15NO
Formula Weight: 177.25
Storage Temperature: 2°C to 8°C
Melting Point: 71-75°
Beilstein Reference: 2044028
MDL No.: MFCD00008799
Formula:
C₁₁H₁₅NO Melting Pt: 71…75 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008799 CAS Number: 3376-24-7 EINECS: 222-168-6 Merck Index: 14,7056 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...