Lithium formate monohydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Formic Acid Lithium Salt, Lithium formate hydrate

A17432-18 A17432-30
CAAAA17432-18EA 61.62 CAD
CAAAA17432-18 CAAAA17432-30
Lithium formate monohydrate 98%
Lithium formate monohydrate

Purity: 98% CAS No.: 6108-23-2 Molecular Formula: LiOOCH.xH2O Formula Weight: 51.96(anhy) Odor: Mild Melting Point: 94° dec. MDL No.: MFCD00050823

Formula: HCO₂Li . H₂O
Melting Pt: 94 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00050823
CAS Number: 6108-23-2
EINECS: 209-133-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR